Sulfuric acid 93-98%, OmniTrace Ultra™ for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

SX1248-1 SX1248-6 SX1248-7
CASX1248-1EA 1361 CAD
CASX1248-1 CASX1248-6 CASX1248-7
Sulfuric acid 93-98%, OmniTrace Ultra™ for trace metal analysis, Supelco®
Sulfuric acid

Clear liquid. For low level trace metal analysis. Double-distilled and packaged in ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom conditions. Supplied in specially designed, preleached fluoropolymer resin bottles to guarantee product integrity. Certificate of Analysis supplied with each shipment.

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830

Order Now

Specification Test Results

Aluminum (Al) 50pg/g max
Antimony (Sb) 50pg/g max
Arsenic (As) 500pg/g max
Assay 93-98%
Barium (Ba) 10pg/g max
Beryllium (Be) 10pg/g max
Bismuth (Bi) 10pg/g max
Cadmium (Cd) 10pg/g max
Calcium (Ca) 50pg/g max
Cerium (Ce) 10pg/g max
Cesium (Cs) 10pg/g max
Chromium (Cr) 10pg/g max
Cobalt (Co) 10pg/g max
Copper (Cu) 10pg/g max
Dysprosium (Dy) 10pg/g max
Erbium (Er) 10pg/g max
Europium (Eu) 10pg/g max
Gadolinium (Gd) 10pg/g max
Gallium (Ga) 10pg/g max
Germanium (Ge) 100pg/g max
Hafnium (Hf) 10pg/g max
Holmium (Ho) 10pg/g max
Indium (In) 10pg/g max
Iron (Fe) 50pg/g max
Lanthanum (La) 10pg/g max
Lead (Pb) 10pg/g max
Lithium (Li) 10pg/g max
Lutetium (Lu) 10pg/g max
Magnesium (Mg) 50pg/g max
Manganese (Mn) 10pg/g max
Mercury (Hg) 100pg/g max
Molybdenum (Mo) 10pg/g max
Neodymium (Nd) 10pg/g max
Nickel (Ni) 50pg/g max
Niobium (Nb) 10pg/g max
Potassium (K) 50pg/g max
Praseodymium (Pr) 10pg/g max
Rhodium (Rh) 50pg/g max
Rubidium (Rb) 10pg/g max
Samarium (Sm) 10pg/g max
Scandium (Sc) 10pg/g max
Selenium (Se) 500pg/g max
Silver (Ag) 50pg/g max
Sodium (N/A) 50pg/g max
Strontium (Sr) 10pg/g max
Tellurium (Te) 50pg/g max
Terbium (Tb) 10pg/g max
Thallium (Tl) 10pg/g max
Thorium (Th) 10pg/g max
Thulium (Tm) 10pg/g max
Tin (Sn) 50pg/g max
Titanium (Ti) 50pg/g max
Tungsten (W) 10pg/g max
Uranium (U) 10pg/g max
VaN/Adium (V) 10pg/g max
Ytterbium (Yb) 10pg/g max
Yttrium (Y) 10pg/g max
Zinc (Zn) 50pg/g max
Zirconium (Zr) 10pg/g max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR