Sulfuric acid 95.5-96.5 %, OmniTrace® for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

SX1247-1 SX1247-2
CASX1247-1CS 1397 CAD
CASX1247-1 CASX1247-2
Sulfuric acid 95.5-96.5 %, OmniTrace® for trace metal analysis, Supelco®
Sulfuric acid

For elemental metal analysis via Atomic Absorption and ICP instruments. Actual Lot Analysis on label. Color-coded label to match acid cap.

Formula: H₂SO₄
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,
Merck Index: 14,08974

Order Now

Specification Test Results

Aluminum (Al) 1ppb max
Antimony (Sb) 1ppb max
Arsenic (As) 0.5ppb max
Assay 93-98%
Barium (Ba) 0.1ppb max
Beryllium (Be) 0.1ppb max
Bismuth (Bi) 0.1ppb max
Cadmium (Cd) 0.5ppb max
Calcium (Ca) 1ppb max
Cerium (Ce) 0.1ppb max
Cesium (Cs) 0.1ppb max
Chloride (Cl) 0.7ppm max
Chromium (Cr) 0.5ppb max
Cobalt (Co) 0.5ppb max
Color (APHA) 10 max
Copper (Cu) 0.5ppb max
Density at 25°C 1.83-1.84g/mL
Dysprosium (Dy) 0.1ppb max
Erbium (Er) 0.1ppb max
Europium (Eu) 0.1ppb max
Gadolinium (Gd) 0.1ppb max
Gallium (Ga) 0.1ppb max
Germanium (Ge) 1ppb max
Gold (Au) 0.5ppb max
Hafnium (Hf) 0.1ppb max
Holmium (Ho) 0.1ppb max
Indium (In) 0.1ppb max
Iron (Fe) 1ppb max
Lanthanum (La) 0.1ppb max
Lead (Pb) 0.1ppb max
Lithium (Li) 0.5ppb max
Lutetium (Lu) 0.1ppb max
Magnesium (Mg) 1ppb max
Manganese (Mn) 0.5ppb max
Mercury (Hg) 0.1ppb max
Molybdenum (Mo) 0.5ppb max
Neodymium (Nd) 0.1ppb max
Nickel (Ni) 0.5ppb max
Niobium (Nb) 0.1ppb max
Nitrate (NO3) 0.2ppm max
Potassium (K) 1ppb max
Praseodymium (Pr) 0.1ppb max
Rhodium (Rh) 0.5ppb max
Rubidium (Rb) 0.5ppb max
Samarium (Sm) 0.1ppb max
Scandium (Sc) 0.1ppb max
Selenium (Se) 10ppb max
Silver (Ag) 1ppb max
Sodium (N/A) 1ppb max
Strontium (Sr) 0.5ppb max
Substances reducing permangaN/Ate 20ppm max
Tellurium (Te) 0.1ppb max
Terbium (Tb) 0.1ppb max
Thallium (Tl) 0.1ppb max
Thorium (Th) 0.1ppb max
Thulium (Tm) 0.1ppb max
Tin (Sn) 1ppb max
Titanium (Ti) 1ppb max
Total phosphorus (P) 0.05ppm max
Tungsten (W) 0.5ppb max
Uranium (U) 0.1ppb max
VaN/Adium (V) 0.5ppb max
Ytterbium (Yb) 0.1ppb max
Yttrium (Y) 0.1ppb max
Zinc (Zn) 1ppb max
Zirconium (Zr) 0.5ppb max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR