KIMAX® Volumetric Flasks with [ST] Glass Stopper, Class B, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
28015-10 28015-100 28015-1000 28015-200 28015-2000 28015-250 28015-50 28015-500
89001-984CS 884.99 CAD
89001-984 89001-986 89001-988 89001-990 89001-992 89001-994 89001-996 89001-998 89002-002
KIMAX® Volumetric Flasks with [ST] Glass Stopper, Class B, Kimble Chase
Flasks Volumetric Flasks
Calibrated to contain
Designed from ASTM specification E288, Class B requirements.
Flasks have a graduation ring blasted on the neck, a marking spot, and a [ST] ground glass stopper.