D-(+)-Sucrose, GR ACS, Sigma-Aldrich®

Supplier: MilliporeSigma

SX1075-20 SX1075-7 SX1075-3
CASX1075-20EA 2978.54 CAD
CASX1075-20 CASX1075-7 CASX1075-1 CASX1075-3
D-(+)-Sucrose, GR ACS, Sigma-Aldrich®
D(+)-Sucrose

Meets ACS Specifications


Certifications: Meets ACS Specifications.

Formula: C₁₂H₂₂O₁₁
Melting Pt: 185…187 °C (lit.)
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride (Cl) 0.005% max
Color White
Form Crystals
Heavy metals (as Pb) 5ppm max
Insoluble matter 0.005% max
Invert sugar 0.05% max
Iron (Fe) 5ppm max
Loss on drying (105°C) 0.03% max
Residue after ignition 0.01% max
Specific rotation [a]25/D +66.3-+66.8°
Sulfate and sulfite (as SO4) 0.005% max
Titrable acid 0.0008meq/g max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR