PYREX® Volumetric Flasks with [ST] Stopper, Corning
Supplier: Corning
5641-10 5641-2L 5641-200 5641-500 5641-50
89091-108CS 928.85 CAD
89091-108 89091-288 89090-608 89090-682 89090-754 89090-772 89090-822 89090-916
PYREX® Volumetric Flasks with [ST] Stopper, Corning
Flasks Volumetric Flasks
Made of PYREX® borosilicate glass.
  • Graduation line is sharp and permanent
  • Large white block letters make inscriptions easy to read
  • Supplied with interchangeable ground glass stoppers
Flasks are designed for those that do not require the precision accuracy of our Class A capacity flasks. Capacity tolerances are twice those specified for class A volumetric ware.