Sodium sulfite, anhydrous ≥98.0%, GR ACS, Supelco®

Supplier: MilliporeSigma

SX0785-5 SX0785-1 SX0785-3
CASX0785-5EA 551 CAD
CASX0785-5 CASX0785-1 CASX0785-3
Sodium sulfite, anhydrous ≥98.0%, GR ACS, Supelco®
Sodium sulphite

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: >500 °C (dec.)
Density: 2.63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 13,08757

Order Now

Specification Test Results

Assay (iodometric) 98.0% min
Chloride (Cl) 0.02% max
Free acid To pass test
Heavy metals (as Pb) 0.001% max
Insoluble matter 0.005% max
Iron (Fe) 0.001% max
Titrable free base 0.03meq/g max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR