Sodium sulfate, anhydrous ≥99.0%, granular, Tracepur® 10-60 mesh, Supelco®

Supplier: MilliporeSigma

Synonyms: Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt

SX0760E-1 SX0760E-20 SX0760E-3 SX0760E-5
CASX0760E-1CS 557 CAD
CASX0760E-1 CASX0760E-20 CASX0760E-3 CASX0760E-5
Sodium sulfate, anhydrous ≥99.0%, granular, Tracepur® 10-60 mesh, Supelco®
Sodium sulphate

Meets ACS Specifications

Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429
Melting Pt: 884 °C (lit.)
Density: 2.68 g/ml (25°C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min
Calcium (Ca) 0.01% max
Chloride (Cl) 0.001% max
Color White
Form Granules
Heavy metals (as Pb) 5ppm max
Insoluble matter 0.01% max
Iron (Fe) 0.001% max
Loss on ignition 0.5% max
Magnesium (Mg) 0.005% max
Mesh size (10-30) 80% min
Mesh size (10-60) 95% min
Mesh size (100 and finer) 1.0% max
Nitrogen compounds (as N) 5ppm max
pH of a 5% solution at 25°C 5.2-9.2
Phosphate (PO4) 0.001% max
Potassium (K) 0.01% max
Suitability for residue analysis (dried 400°/4 hr) To pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR