Sodium nitrite ≥97.0%, GR ACS, Supelco®

Supplier: MilliporeSigma

Synonyms: Diazotizing salts

SX0665-1 SX0665-7 SX0665-7 SX0665-1
CASX0665-1CS 634.31 CAD
EM-SX0665-1 CASX0665-1 EM-SX0665-7 CA-SX0665-7
Sodium nitrite ≥97.0%, GR ACS, Supelco®
Sodium nitrite

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: NaNO₂
MW: 69 g/mol
Melting Pt: 280 °C (dec.)
MDL Number: MFCD00011118
CAS Number: 7632-00-0
UN: 1500
ADR: 5.1,6.1,III
Merck Index: 13,08721

Order Now

Specification Test Results

Assay 99.0% min
Calcium (Ca) 0.002% max
Chloride (Cl) 0.005% max
Heavy metals (as Pb) 0.001% max
Insoluble matter 0.01% max
Iron (Fe) 0.001% max
Potassium (K) 0.001% max
Sulfate (SO4) 0.005% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR