Sodium hexametaphosphate ≥65%, Supelco®

Supplier: MilliporeSigma

Synonyms: Sodium metaphosphateSHMP

SX0583-3 SX0583-3
CASX0583-3EA 484.09 CAD
CASX0583-3 EM-SX0583-3
Sodium hexametaphosphate ≥65%, Supelco®
Sodium hexametaphosphate

Granular.

Formula: Na₆P₆O₁₈
MW: 611.77 g/mol
Melting Pt: 610 °C
Density: 1.25 g/cm³ (20 °C)
MDL Number: MFCD00136045
CAS Number: 10124-56-8

Order Now

Specification Test Results

Assay (as P2 O5) 65% min
Color White
Form Granules

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR