tri-Sodium citrate dihydrate 99.0-101.0% (by acidimetry) GR ACS, Supelco®

Supplier: MilliporeSigma

Synonyms: Sodium Citrate, DihydrateCitric acid trisodium salt dihydrateSodium Citrate DihydrateSodium citrate tribasic dihydrateSodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate

SX0445-20 SX0445-20
CA-SX0445-20EA 7445.75 CAD
TXSX044520SRA CA-SX0445-20
tri-Sodium citrate dihydrate 99.0-101.0% (by acidimetry) GR ACS, Supelco®
tri-Sodium citrate dihydrate

Meets ACS Specifications

Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 150 °C (anhydr.)
Density: 1.76 g/cm³ (20 °C)
MDL Number: MFCD00150031
CAS Number: 6132-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR