Orthophosphoric acid ≥85.0-88.0% NF, Multi-Compendial

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Phosphoric acid

Management of Change (MOC) category = R

BAKR0268-00 BAKR0268-03 BAKR0268-06 BAKR0268-05 BAKR0268-07 BAKR0268-15
CAJT0268-0CS 2543.5 CAD
CAJT0268-0 CAJT0268-3 CAJT0268-6 CAJT0268-5 CAJT0268-07 CAJT0268-QM CAJT0268-15
Orthophosphoric acid ≥85.0-88.0% NF, Multi-Compendial
Orthophosphoric acid
Formula: H₃PO₄
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.69 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III
REACH: 01-2119490790-32
Merck Index: 13,08662

Order Now

Specification Test Results

GMP Manufactured Product
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification Passes Test
NF - Nitrate (NO₃) Passes Test
NF - Phosphorous or Hypophosphorous Acid Passes Test
NF - Sulfate (SO₄) Passes Test
NF - Alkali Phosphates Passes Test
NF - Assay 85.0 - 88.0 %
EP/BP - Assay (H₃PO₄) 84.0 - 90.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Substances Precipitated with Ammonia Passes Test
EP/BP - Phosphorous or Hypophosphorous Acid Passes Test
EP/BP - Chloride (Cl) ≤ 50 ppm
EP/BP - Sulfate (SO₄) ≤ 100 ppm
EP/BP - Iron (Fe) ≤ 50 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR