Orthophosphoric acid ≥85.0-88.0% NF, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

BAKR0262-01 BAKR0262-15 BAKR0262-03 BAKR0262-04 BAKR0262-06 BAKR0262-07 BAKR0262-02 BAKR0262-05 BAKR0262-08
CAJT0262-1CS 6258.63 CAD
CAJT0262-1 CAJT0262-15 CAJT0262-3 CAJT0262-4 CAJT0262-6 CAJT0262-7 CAJT0262-2 CAJT0262-5 CAJT0262-08 CAJT0262-QM
Orthophosphoric acid ≥85.0-88.0% NF, FCC, J.T.Baker®
Orthophosphoric acid
Formula: H₃PO₄
Melting Pt: 28 °C
Density: 1.69 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Food GMP Manufactured Product
GMP Manufactured Product
Meets N.F. Requirements
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
NF - Assay (H₃PO₄) 85.0 - 88.0 %
NF - Identification Passes Test
NF - Nitrate (NO₃) Passes Test
NF - Phosphorous or Hypophosphorous Acid Passes Test
NF - Sulfate (SO₄) Passes Test
NF - Alkali Phosphates Passes Test
FCC - Assay (H₃PO₄) 85.0 - 88.0 %
FCC - Identification Passes Test
FCC - Arsenic (As) ≤3 mg/kg
FCC - Cadmium (Cd) ≤3 mg/kg
FCC - Lead (Pb) ≤3 mg/kg
FCC - Fluoride (F) ≤10 mg/kg
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR