Orthophosphoric acid ≥85.0-87.0%, BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

BAKR0260-00 BAKR0260-01 BAKR0260-15 BAKR0260-33 BAKR0260-07 BAKR0260-08 BAKR0260-05 0260-02 0260-03
CAJT0260-0CS 1688.74 CAD
CAJT0260-0 CAJT0260-1 CAJT0260-15 CAJT0260-33 CAJT0260-7 CAJT0260-8 CAJT0260-5 JT0260-2 JT0260-3
Orthophosphoric acid ≥85.0-87.0%, BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®
Orthophosphoric acid

The quality of BAKER ANALYZED™ ACS Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.

Formula: H₃PO₄
Melting Pt: 28 °C
Density: 1.69 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (H₃PO₄) (by acidimetry) 85.0 - 87.0 %
Calcium (Ca) ≤0.002 %
Color (APHA) ≤10
Insoluble Matter ≤0.001 %
ACS - Magnesium (Mg) ≤0.002 %
Sulfate (SO₄) ≤12 ppm
Volatile Acids (as CH₃COOH) ≤0.001 %
Reducing Substances Passes Test
Chloride (Cl) ≤3 ppm
Nitrate (NO₃) ≤5 ppm
Trace Impurities - Antimony (Sb) ≤20.000 ppm
Trace Impurities - Arsenic (As) ≤0.500 ppm
Trace Impurities - Iron (Fe) ≤10 ppm
Heavy Metals (as Pb) ≤8 ppm
Trace Impurities - Manganese (Mn) ≤0.500 ppm
Trace Impurities - Potassium (K) ≤40.000 ppm
Trace Impurities - Sodium (Na) ≤200.000 ppm
Exceeds A.C.S. Specifications
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR