Silica gel 60-200 mesh Grade 62, Supelco®

Supplier: MilliporeSigma

SX0143U-1 SX0143U-3
CASX0143U-1EA 356 CAD
CASX0143U-1 CASX0143U-3
Silica gel 60-200 mesh Grade 62, Supelco®
Silica gel

60 to 200 mesh nominal.

Formula: SiO₂
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
REACH: 01-2119379499-16

Order Now

Specification Test Results

Color White
Form Coarse powder
Mesh size (60-200) 80% min


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR