Silica gel 100-200 mesh Grade 923, Supelco®
Supplier: MilliporeSigma
SX0143L-1
SX0143L-1
CASX0143L-1CS
9087.69
CAD
CASX0143L-1
EM-SX0143L-1
Silica gel 100-200 mesh Grade 923, Supelco®
Silica gel
100 to 200 mesh.
Formula:
SiO₂ MW: 60.08 g/mol Boiling Pt: 2230 °C (1013 hPa) Melting Pt: 1710 °C |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 |
Specification Test Results
Color | White |
Form | Powder |
Mesh size (100 - 200) | To pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...