Sand, quartz (from sea), Supelco®

Supplier: MilliporeSigma

SX0076-30 SX0076-1
CASX0076-30EA 641 CAD
CASX0076-30 CASX0076-1
Sand, quartz (from sea), Supelco®
Sand, quartz

For heating baths.

Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: >2200 °C (1013 hPa)
Melting Pt: 1713 °C
Density: 2.622 to 2.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD02100519
CAS Number: 14808-60-7
EINECS: 238-878-4

Order Now

Specification Test Results

Color Beige
Description To pass test
Form Granules, free-flowing, visibly dry

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR