Sand, quartz standard (Ottawa), Supelco®

Supplier: MilliporeSigma

SX0070-1 SX0070-3
CASX0070-1CS 616 CAD
CASX0070-1 CASX0070-3
Sand, quartz standard (Ottawa), Supelco®
Sand, quartz
Formula: SiO₂
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1427 °C
Density: 2.622…2.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD02100519
CAS Number: 14808-60-7
EINECS: 238-878-4

Order Now

Specification Test Results

Color Beige
Description To pass test
Form Granules


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR