Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 700)

Supplier: AAT BIOQUEST INC
116100J0 116100J1
77023-628EA 385.9 CAD
77023-628 77021-586
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 700)
Antibodies
HP-3G10 is an anti-human monoclonal antibody that recognizes the CD161 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR