Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 860)

Supplier: AAT BIOQUEST INC
104410R0 104410R1
77021-182EA 385.9 CAD
77021-182 77021-552
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 860)
Antibodies
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: iFluor® 860
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR