Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (PE/Cy5)

Supplier: AAT Bioquest
103801M1 103801M0 103801M2
77023-166EA 924.25 CAD
77023-166 77022-068 77023-072
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (PE/Cy5)
Antibodies
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: PE/Cy5
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR