Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 710)

Supplier: AAT BIOQUEST INC
103800K0 103800K1
77021-014EA 385.9 CAD
77021-014 77021-086
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 710)
Antibodies
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: iFluor® 710
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR