D-(+)-Sucrose solution, OmniPur®, Millipore®

Supplier: MilliporeSigma

Synonyms: α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranosideSucroseD(+)-Saccharoseβ-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranosideα-ᴅ-Glc-(1→2)-β-ᴅ-Fru

8590-100ML 8590-100ML
CAEM8590EA 180.32 CAD
CAEM8590 EM-8590
D-(+)-Sucrose solution, OmniPur®, Millipore®
D(+)-Sucrose

Sterile. DNase-, RNase-, and protease-free.

Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
MDL Number: MFCD00006626
CAS Number: 57-50-1
https://ca.vwr.com/stibo/web/std.lang.all/95/86/2299586.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR