D-(+)-Sucrose, OmniPur®, Millipore®

Supplier: MilliporeSigma

8510-500GM 8550-5KG 8510-500GM
CA-EM8510EA 267.05 CAD
CA-EM8510 CA-EM8550 CA76000-908
D-(+)-Sucrose, OmniPur®, Millipore®
D(+)-Sucrose

DNase-, RNase-, and protease-free.

Formula: C₁₂H₂₂O₁₁
Melting Pt: 185…187 °C (lit.)
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR