Sodium dihydrogen phosphate monohydrate 98.0-102.0%, OmniPur® DNase-, RNase-, protease-free, Millipore®

Supplier: MilliporeSigma

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

8290-500GM 8295-5KG
CA-EM8290EA 218.16 CAD
CA-EM8290 CAEM8295
Sodium dihydrogen phosphate monohydrate 98.0-102.0%, OmniPur® DNase-, RNase-, protease-free, Millipore®
Sodium dihydrogen phosphate monohydrate

DNase-, RNase-, and protease-free.

Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
EINECS: 231-449-2
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Arsenic 0.00005% max.
Assay 98.0–102.0%
Chloride 0.0005% max.
Heavy Metals (as Pb) 0.001% max.
Insolubles 0.01% max.
Iron 0.001% max.
Nitrogen Compounds 0.001% max.
pH (5%, H2O) 4.1–4.5
Sulfate 0.003% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR