Disodium hydrogen phosphate heptahydrate ≥98.0%, OmniPur®, Molecular Biology Grade, Millipore®

Supplier: MilliporeSigma

Synonyms: Sodium phosphate dibasic heptahydrate, Sodium hydrogen phosphate heptahydrate, Sodium monohydrogen phosphate heptahydrate

8210-500GM 8240-5KG
CA-EM8210EA 152.47 CAD
CA-EM8210 CAEM8240
Disodium hydrogen phosphate heptahydrate ≥98.0%, OmniPur®, Molecular Biology Grade, Millipore®
di-Sodium hydrogen phosphate heptahydrate

DNase-, RNase-, and protease-free.


Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Formula: HNa₂O₄P·7H₂O
MW: 268.07 g/mol
Melting Pt: 48
Density: 1.68 g/ml (25°C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00149180
CAS Number: 7782-85-6
EINECS: 231-448-7

Order Now

Specification Test Results

Arsenic (As) 0.0005% max
Assay 98.0% min
Chloride (Cl) 0.001% max
DNase None detected
Heavy metals (as Pb) 0.001% max
Insoluble matter 0.005% max
Iron (Fe) 0.001% max
Moisture (H2O) 43-50%
Nitrogen compounds (as N) 0.001% max
pH (5%, water) 8.7-9.3
Protease None detected
RNase None detected
Sulfate (SO4) 0.005% max 


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR