Sodium dodecyl sulfate (SDS) ≥99.0%, OmniPur®, Millipore®

Supplier: MilliporeSigma
Danger

7910-500GM 7960-5KG
CA-EM7910EA 826.34 CAD
CA-EM7910 CA-EM7960
Sodium dodecyl sulfate (SDS) ≥99.0%, OmniPur®, Millipore®
Sodium dodecyl sulphate (SDS)

DNase-, RNase-, protease-, and phosphatase-free.

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR