Anti-CD28 Mouse Monoclonal Antibody (mFluor™ Violet) [clone: CD28.2]

Supplier: AAT BIOQUEST INC
10280120 10280121
77022-442EA 385.9 CAD
77022-442 77021-362
Anti-CD28 Mouse Monoclonal Antibody (mFluor™ Violet) [clone: CD28.2]
Antibodies
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: mFluor™ Violet
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR