Anti-CD28 Mouse Monoclonal Antibody (iFluor® 700) [clone: CD28.2]

Supplier: AAT BIOQUEST INC
102800J0 102800J1
77024-924EA 385.9 CAD
77024-924 77021-010
Anti-CD28 Mouse Monoclonal Antibody (iFluor® 700) [clone: CD28.2]
Antibodies
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD28
Antigen name CD28 antigen
Environmentally Preferable
Antibody type Secondary
Clonality Monoclonal
Conjugation iFluor® 700
Clone CD28.2
Reactivity Human
Host Mouse
Isotype
Purification Affinity purification
Storage temperature Refrigerated (2...8 °C); minimize light exposure
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR