Anti-CD19 Mouse Monoclonal Antibody (PerCP/Cy5.5) [clone: SJ25C1]

Supplier: AAT BIOQUEST INC
101911U0 101911U1 101911U2
77021-034EA 332.91 CAD
77021-034 77021-170 77021-688
Anti-CD19 Mouse Monoclonal Antibody (PerCP/Cy5.5) [clone: SJ25C1]
Antibodies
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: PerCP/Cy5.5
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD19
Antigen name CD19 antigen
Environmentally Preferable
Antibody type Secondary
Clonality Monoclonal
Conjugation PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Clone SJ25C1
Reactivity Human
Host Mouse
Isotype
Purification Affinity purification
Storage temperature Refrigerated (2...8 °C); minimize light exposure
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR