Anti-CD19 Mouse Monoclonal Antibody (iFluor® 568) [clone: SJ25C1]

Supplier: AAT BIOQUEST INC
101910B0 101910B1
77023-120EA 385.9 CAD
77023-120 77024-776
Anti-CD19 Mouse Monoclonal Antibody (iFluor® 568) [clone: SJ25C1]
Antibodies
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: iFluor® 568
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR