Anti-CD14 Mouse Monoclonal Antibody (PE/iFluor® 700) [clone: 61D3]

Supplier: AAT BIOQUEST INC
101411X0 101411X1 101411X2
77022-702EA 332.91 CAD
77022-702 77023-754 77023-524
Anti-CD14 Mouse Monoclonal Antibody (PE/iFluor® 700) [clone: 61D3]
Antibodies
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: PE/iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR