Anti-CD14 Mouse Monoclonal Antibody (iFluor® 488) [clone: 61D3]

Supplier: AAT BIOQUEST INC
10141050 10141051
77022-756EA 385.9 CAD
77022-756 77023-902
Anti-CD14 Mouse Monoclonal Antibody (iFluor® 488) [clone: 61D3]
Antibodies
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: iFluor® 488
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD14
Antigen name CD14 antigen
Environmentally Preferable
Antibody type Secondary
Clonality Monoclonal
Conjugation iFluor® 488
Clone 61D3
Reactivity Human
Host Mouse
Isotype
Purification Affinity purification
Storage temperature Refrigerated (2...8 °C); minimize light exposure
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR