Anti-CD8a Mouse Monoclonal Antibody (iFluor® A7) [clone: SK1]

Supplier: AAT BIOQUEST INC
100810S0 100810S1
77024-222EA 385.9 CAD
77024-222 77023-596
Anti-CD8a Mouse Monoclonal Antibody (iFluor® A7) [clone: SK1]
Antibodies
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: iFluor® A7
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR