Anti-CD8a Mouse Monoclonal Antibody (iFluor® 610) [clone: SK1]

Supplier: AAT BIOQUEST INC
100810D0 100810D1
77024-512EA 385.9 CAD
77024-512 77021-726
Anti-CD8a Mouse Monoclonal Antibody (iFluor® 610) [clone: SK1]
Antibodies
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: iFluor® 610
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR