Anti-CD5 Mouse Monoclonal Antibody (iFluor® 800) [clone: UCHT2]

Supplier: AAT BIOQUEST INC
100520N0 100520N1
77023-416EA 385.9 CAD
77023-416 77022-998
Anti-CD5 Mouse Monoclonal Antibody (iFluor® 800) [clone: UCHT2]
Antibodies
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: iFluor® 800
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR