Anti-CD5 Mouse Monoclonal Antibody (PE/iFluor® 750) [clone: L17F12]

Supplier: AAT BIOQUEST INC
100511R0 100511R1 100511R2
77022-866EA 332.91 CAD
77022-866 77024-732 77021-528
Anti-CD5 Mouse Monoclonal Antibody (PE/iFluor® 750) [clone: L17F12]
Antibodies
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: PE/iFluor® 750
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR