PYREXPLUS® Plastic-Coated Volumetric Flasks, [ST] Joint, Class A, Corning
Supplier: Corning
65640-100 65640-1L 65640-2L 65640-500
29618-751CS 2377.43 CAD
29618-751 89091-674 89091-678 89091-688 89091-692
PYREXPLUS® Plastic-Coated Volumetric Flasks, [ST] Joint, Class A, Corning
Flasks Volumetric Flasks
PYREX® borosilicate glass, plastic coated, clear, with glass stoppers.
  • Resistant to thermal shock
  • Strength increased through machine-blown bodies
  • Sharp graduation line and permanent and large white block letters make the inscription easy to read
These flasks features a protective PVC coating for safety and long life, and it prevents glass from shattering and reduces spills.
Can be written on with any standard lab marker, permanent or water-soluble. Supplied with [ST] PYREX® stopper. It allows for a better hand grip and clamp hold.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Monterrey