Sodium chloride ≥99.5%, OmniPur®, Millipore®
Supplier: MilliporeSigma
7710-500GM
7760-5KG
7780-25KG
CA-EM7710EA
529.14
CAD
CA-EM7710
CA-EM7760
CAEM7780
Sodium chloride ≥99.5%, OmniPur®, Millipore®
Sodium chloride
DNase-, RNase-, protease-, and phosphatase-free.
Formula:
NaCl Boiling Pt: 1413 °C (1013 hPa) Melting Pt: 801 °C (lit.) Density: 2.16 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,08599 |
Specification Test Results
Assay | 99.5% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...