VWR® TraceClean® Leakproof Wide Mouth Containers, HDPE
Supplier: VWR International
76184-820 76184-822 76184-824 76184-826 76184-828 76184-830 76184-832 76184-834 76184-836 76184-838 76184-840 76184-842 76184-844 76184-846 76184-848 76184-850 76184-852 76184-854 76184-856 76184-858
76184-820CS 118.86 CAD
76184-820 76184-822 76184-824 76184-826 76184-828 76184-830 76184-832 76184-834 76184-836 76184-838 76184-840 76184-842 76184-844 76184-846 76184-848 76184-850 76184-852 76184-854 76184-856 76184-858
VWR® TraceClean® Leakproof Wide Mouth Containers, HDPE
Bottles Water Sampling Bottles
TraceClean® containers are used primarily for the collection of aqueous media for testing metals and a variety of inorganic parameters including alkalinity, anions, acidity, cyanide, sulfate, and hardness. Not all analytical parameters are included on the Certificate of Analysis. Please refer to the Certificate of Analysis for certified analytes and corresponding reporting limits. Secondary uses include containment and archiving.
Quality Assured® with VWR® Logo Sample ID Labels (QAL) containing the Certificate of Analysis and Cleaning Protocol Form in the product case.
Quality Assured®More Product Information