Potassium dihydrogen phosphate, crystallised, EMPROVE® ESSENTIAL JPC, NF, Ph. Eur., BP, E340, SAFC®

Supplier: EMD MILLIPORE – PCS CA EXCEP

Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

1.04871.5000 1.04871.9029 1.04871.9050
EM1.04871.5000EA 1306.72 CAD
EM1.04871.5000 CA1.04871.9029 CA1.04871.9050
Potassium dihydrogen phosphate, crystallised, EMPROVE® ESSENTIAL JPC, NF, Ph. Eur., BP, E340, SAFC®
Potassium dihydrogen phosphate

Crystals.


Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Certifications: Corresponds to Ph. Eur., BP, JPC, NF, E 340
Conforms to the purity criteria on food additives according to the current European Commission Regulation.

Formula: KH₂PO₄
MW: 136.09 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 253
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Order Now

Specification Test Results

Assay (alkalimetric, calculated on dried substance) 98.0 - 100.5 %
Assay (alkalimetric; dried substance) ≥ 99.0 %
Appearance of solution passes test
Identity passes test
Insoluble substances ≤ 0.2 %
pH-value (1 %; water) 4.2 - 4.8
pH-value (2 %; water) 4.2 - 4.6
pH-value (5 %; water) 4.2 - 4.5
Carbonate passes test
Condensed phosphate passes test
Chloride (Cl) ≤ 0.009 %
Fluoride (F) ≤ 0.0010 %
Sulfate (SO₄) ≤ 0.017 %
Heavy metals (as Pb) ≤ 0.0010 %
Al (Aluminium) ≤ 0.0005 %
As (Arsenic) ≤ 0.0001 %
Cd (Cadmium) ≤ 0.0001 %
Fe (Iron) ≤ 0.0010 %
Hg (Mercury) ≤ 0.0001 %
Na (Sodium) passes test
Pb (Lead) ≤ 0.0001 %
Residual solvents (ICH Q3C) excluded by manufacturing process
Reducing substances passes test
Loss on drying (105 °C) ≤ 0.5 %
Loss on drying (130 °C) ≤ 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR