Pyridine, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Azabenzene, Azine

PX2020-11 PX2020-5 PX2020-6
CAPX2020-11CS 2310 CAD
CAPX2020-11 CAPX2020-5 CAPX2020-6
Pyridine, GR ACS, Supelco®
Pyridine

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115
Melting Pt: -44
Density: 0.983 g/cm³ (20 °C)
Flash Pt: 17
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3 II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.0% min
Ammonia (NH3) 0.002% max
Chloride (Cl) 0.001% max
Copper (about 5 ppm) To pass test
Form Clear liquid
Reducing substances To pass test
Residue after evaporation 0.002% max
Solubility in water To pass test
Sulfate (SO4) 0.001% max
Water (H2O) 0.1% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR