Pyridine, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

PX2014-1 PX2014-6 PX2014-6
CAPX2014-1EA 1834 CAD
CAPX2014-1 CAPX2014-6 EM-PX2014-6
Pyridine, OmniSolv®, Supelco®
Pyridine

Suitable for spectrophotometry and HPLC and GC. UV cutoff at 330nm. Filtered to remove particulate matter. Lot analysis on label.

Formula: C₅H₅N
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.983 g/cm³ (20 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.0% min
Color (APHA) at time of manufacture 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 4ppm max
UV Abs. at 330 nm 1.00AU max
UV Abs. at 340 nm 0.30AU max
UV Abs. at 350 nm 0.10AU max
UV Abs. at 400 nm 0.01AU max
UV Cut-off 330nm max
Water (H2O) 0.05% max

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR