Butyl Synthetic Rubber Gloves, Showa
Supplier: Showa-Best Glove
878-09 878-10 878-11 878-08
CA32892-050PK 147.47 CAD
CA32892-050 CA32892-051 CA32892-052 CA47727-168
Butyl Synthetic Rubber Gloves, Showa
Gloves
Highly resistant to corrosive acids.
  • Excellent hand protection for handling esters and ketones
  • Provide the highest permeation resistance against gases and water vapor of any glove material
Each pair of injection-molded, hand-specific gloves are nonpowdered, unlined, and unsupported. Cuff is rolled. Smooth, 30 mil butyl is flexible and durable, but not as strong as natural rubber. It provides good chemical resistance to bases, alcohols, amines and amides, glycol esters, nitrocompounds, and aldehydes, but does not perform well with halogen compounds, aliphatic hydrocarbons, or aromatic hydrocarbons. Color: black.