Round-Bottom Three-Neck Flasks, Vertical Necks, Socket Joints, Ace Glass
Supplier: Ace Glass
6950-08 6950-208 6950-222 6950-230 6950-238 6950-244 6950-250 6950-30 6950-38 6950-44 6950-50 6950-60 6950-22 6950-54
89056-934EA 410.45 CAD
89056-934 89056-936 89056-938 89056-940 89056-942 89056-944 89056-946 89056-948 89056-950 89056-952 89056-954 89056-956 89083-884 89083-906
Round-Bottom Three-Neck Flasks, Vertical Necks, Socket Joints, Ace Glass
Flasks Round Bottom Flasks
Round-bottom flasks with three vertical necks and [SJ] socket joints
Heavy wall flasks feature walls that are approximately 30% heavier than standard flasks. Side necks all have [SJ] 35/25 socket joints.