Round-Bottom Flasks with Beaded Side Arm, Ace Glass
Supplier: Ace Glass
6935-03 6935-04 6935-05 6935-09 6935-13 6935-17 9460-02 9460-06 9460-08 9460-04 9460-10 9460-12
89056-522EA 143.12 CAD
89056-522 89056-524 89056-526 89056-528 89056-530 89056-532 89074-644 89074-650 89074-652 89074-834 89074-878 89074-880
Round-Bottom Flasks with Beaded Side Arm, Ace Glass
Flasks Round Bottom Flasks
This round-bottom flask has a center neck with [ST] outer joint and beaded side port
The outer joint is tooled with reinforcement rings (full sized joints only) for added strength and stability. The I.D. of the side port is approximately 9.5 mm. Fabricated from borosilicate glass.
Ordering information: For a version of this flask with a deep side well rather than a side port, see For a version of this flask with a deep side well rather than a side port, see 89056-534.