Pear-Shaped Drying Flasks, Side Indents, Heavy Wall, Ace Glass
Supplier: Ace Glass
3994-22 3994-10 3994-12 3994-14 3994-20 3994-23 3994-124 3994-126 3994-120 3994-112 3994-114 3994-110
89083-030EA 407.03 CAD
89083-030 89083-032 89083-034 89083-036 89083-044 89083-046 89083-052 89083-054 89083-056 89083-076 89083-078 89083-080
Pear-Shaped Drying Flasks, Side Indents, Heavy Wall, Ace Glass
Flasks Round Bottom Flasks
Pear-shaped evaporating flasks with a [ST] outer joint and side indents
Both uncoated and poly-coated flasks are suitable for rotary evaporators. The outer joint is tooled with reinforcement rings for added strength and stability. Heavy wall flasks feature walls that are approximately 30% heavier than standard flasks. Fabricated from borosilicate glass.