Flask, Tapered Wall, Five Necks, Two #7 Ace-Threds, Ace Glass Incorporated
Supplier: Ace Glass
6963-17 6963-19 6963-60 9473-15 9473-13
89057-292EA 716.29 CAD
89057-292 89057-294 89057-300 89074-858 89074-860
Flask, Tapered Wall, Five Necks, Two #7 Ace-Threds, Ace Glass Incorporated
Flasks Round Bottom Flasks
This flask has walls tapered inward toward the bottom
This type design allows continuous operation with smaller volumes. Supplied with a standard taper outer center neck, two standard taper outer angled side necks, and two #7 Ace-Thred ports for thermometers, miniature electrodes, etc., up to 7mm O.D. Full sized joints (only) are tooled with reinforcement rings for added strength and stability. Fabricated from borosilicate glass.
Ordering information: Supplied complete with nylon bushings and FETFE o-rings.
Order Now
SPECIFICATIONS
Environmentally Preferable