Flask, Tapered Wall, Four Necks, One #7 Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
6963-34 6963-36 9473-20 9473-24
89057-288EA 655.67 CAD
89057-288 89057-290 89074-848 89074-854
Flask, Tapered Wall, Four Necks, One #7 Ace-Thred, Ace Glass Incorporated
Flasks Round Bottom Flasks
This flask has walls tapered inward toward the bottom
This type design allows continuous operation with smaller volumes. Supplied with a standard taper outer center neck, two standard taper outer angled side necks, and one #7 Ace-Thred port for thermometers, miniature electrodes, etc., up to 7mm O.D. Full sized joints (only) are tooled with reinforcement rings for added strength and stability. Fabricated from borosilicate glass.
Ordering information: Supplied complete with nylon bushing and FETFE o-ring.
Order Now
SPECIFICATIONS
Environmentally Preferable