European Style Three-Neck Flasks, Angled Necks, Ace Glass
Supplier: Ace Glass
6961-04 6961-09 6961-16
89057-258EA 415.23 CAD
89057-258 89057-260 89057-262
European Style Three-Neck Flasks, Angled Necks, Ace Glass
Flasks Round Bottom Flasks
European style flasks have tapered walls with a shallow bottom for more efficient magnetic or mechanical half-moon shaped paddle stirring
The vertical center neck and angled side necks have [ST] outer joints that are tooled with reinforcement rings for added strength and stability. Fabricated from borosilicate glass.