European Style Four-Neck Flasks, Angled Necks, Ace Glass
Supplier: Ace Glass
6961-39 6961-40 6961-45 6961-34
89057-266EA 541.65 CAD
89057-266 89057-268 89057-270 89057-272
European Style Four-Neck Flasks, Angled Necks, Ace Glass
Flasks Round Bottom Flasks
European style flasks have tapered walls with a shallow bottom for more efficient magnetic or mechanical half-moon shaped paddle stirring
These four-neck flasks have three [ST] outer joints and #7 Ace-Thred neck. The outer joints are tooled with reinforcement rings for added strength and stability. Two of the side necks are in-line with the center neck. The remaining side neck is placed in front of the in-line necks and is internally threaded to accommodate thermometers or bleed tubes up to 7mmO.D. Supplied with #7 nylon bushing and FETFE O-ring. Fabricated from borosilicate glass.